Brewers yeast, MP Biomedicals
Supplier: MP Biomedicals
0290331205 0290331225 0290331280
ICNA0290331205EA 311 SGD
ICNA0290331205 ICNA0290331225 ICNA0290331280
Brewers yeast, MP Biomedicals
Cell Culture Supplements and Additives
Brewers yeast is often used as a source of B-complex vitamins, chromium, and selenium. It is also can be used to bake bread, ferment wine, beer and ethanol production.