Vitamin mixture, AIN-76-A, MP Biomedicals
Supplier: MP Biomedicals
0296009801
ICNA0296009801EA 1330 SGD
ICNA0296009801
Vitamin mixture, AIN-76-A, MP Biomedicals
Animal Supplies Animal Feed
Basic component of animal diet especially for maintaining mice and rats colonies in the research laboratory.
Please see product datasheet for complete diet composition.
Delivery information: In powdered form.
Order Now
SPECIFICATIONS
Environmentally Preferable