Vitamin mixture, AIN-93-VX, MP Biomedicals
Supplier: MP Biomedicals
0296040201
ICNA0296040201EA 409 SGD
ICNA0296040201
Vitamin mixture, AIN-93-VX, MP Biomedicals
Animal Supplies Animal Feed
AIN-93-VX vitamin mixture is a component of AIN-93M and AIN-93G diet.
This vitamin mix is used for the rodents diet revised after numerous nutritional and technical problems were encountered, especially during growth, pregnancy, and lactation.
Caution: Not available in pellet form.