Water cell culture reagent DNase-, RNase-free

Supplier: MP Biomedicals

04821739
ICNA04821739EA 179 SGD
ICNA04821739
Water cell culture reagent DNase-, RNase-free
Water

Deionized water treated with 0.001% diethylpyrocarbonate (DEPC). Filtered through 0.2 micron filter and autoclaved to yield a sterile solution completely free of detectable DNase (both exo and endo) and RNase.


  • DNase, RNase-free

Formula: H₂O
Storage Temperature: Ambient
MDL Number: MFCD00011332
CAS Number: 7732-18-5

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR