2-(2-Aminoethyl)pyridine

Supplier: MP Biomedicals

0520936910
ICNA0520936910EA 472 SGD
ICNA0520936910
2-(2-Aminoethyl)pyridine
2-(2-Aminoethyl)pyridine
Formula: C₇H₁₀N₂
Boiling Pt: 49…50 °C (0,03 mmHg)
Density: 1,02 g/cm³ (20 °C)
Storage Temperature: Ambient
MDL Number: MFCD00006367
CAS Number: 2706-56-1
EINECS: 220-295-1

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR