Streptomyces hyalurolyticus hyaluronidase, MP Biomedicals
Supplier: MP Biomedicals
08320421
ICNA08320421EA 1410 SGD
ICNA08320421
Streptomyces hyalurolyticus hyaluronidase, MP Biomedicals
Enzymes
The hyaluronidase is purified from Streptomyces hyalurolyticus. This enzyme belongs to the endo-beta-hexosaminidase and endo-eliminase type enzyme that produces oligo-saccharide (unsaturated tetraose and pentose) by hydrolysing the beta-1,4-linkage of hyaluronic acid. It is stable in both high temperature and acid or alkaline range. It has high substrate specificity and hydrolyzes hyaluronic acid only in mucopolysaccharide.
  • Inhibitors: Mn++, Mg++, potassium ferricyanide
  • Free from protease and lysozyme
Unit definition: One unit is defined as the quantity of enzyme that causes 50% of decrease in absorbance at 660 nm in 30 minutes at 60 °C.