Aspergillus japonicus Pectolyase Y-23, MP Biomedicals

Supplier: MP Biomedicals

08320951
ICNA08320951EA 740 SGD
ICNA08320951
Aspergillus japonicus Pectolyase Y-23, MP Biomedicals
Enzymes

Pectolyase Y-23 contains two types of pectinase such as endopolygalacturonase and endo-pectin lyase in high activity. It includes maceration stimulating factor which stimulates tissue maceration by both pectinases.


  • Soluble in water


It is used for isolation of biologically active protoplasts from a wide spectrum of higher plants and tissues in combination with a cellulase preparation in a brief incubation.


Unit definition: One unit is defined as an increase in A235 by 1,0 in the reaction mixture per minute.

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR