Anti-Albumin Goat Polyclonal Antibody

Supplier: MP Biomedicals
0855028
ICNA0855028EA 940 SGD
ICNA0855028
Anti-Albumin Goat Polyclonal Antibody
Antibodies
The IgG fraction is prepared from the specific goat antiserum by delipidation, ammonium sulfate fractionation and anion exchange chromatography. The resulting IgG fraction is dialyzed into 0.02M sodium phosphate, 0.14M sodium chloride, pH 7.3, with 0.05% sodium azide, adjusted to standard titer, filtered through a 0.22 µm filter, vialed and lyophilized

The total protein is measured using the Biuret procedure, with bovine albumin as a standard. Antibody titer is standardized to an in-house standard by equivalence-point precipitation. Each IgG fraction is tested for purity and specificity at aMore Product Information
Order Now

SPECIFICATIONS

Antigen name Albumin
Environmentally Preferable
Antibody type Primary
Clonality Polyclonal
Conjugation Unconjugated
Reactivity Human
Host Goat
Isotype IgG
Immunogen Human albumin
Form Lyophilized
Purification Delipidation, ammonium sulfate fractionation, and anion exchange chromatography
Concentration Total Protein: 30.8 mg/mL; Antibody Protein: 8 mg/mL

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR