Anti-kappa chain (bound) Antibody

Supplier: MP Biomedicals
0855059
ICNA0855059EA 1260 SGD
ICNA0855059
Anti-kappa chain (bound) Antibody
Antibodies
The total protein is measured using the Biuret procedure, with bovine albumin as a standard. Antibody titer of the IgG fraction is standardized by equivalence-point precipitation. F(ab')2 fragments are tested for purity and specificity at a minimum of 20 mg/mL using immunoelectrophoresis. The product is goat F(ab')2; no trace of albumin, whole molecule IgG or Fc is detected.

Type: Secondary
Antigen: Kappa Chain (bound)
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Goat
Isotype: IgG
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR