Anti-Ig Goat Antibody (FITC (Fluorescein Isothiocyanate))

Supplier: MP Biomedicals
0855499
ICNA0855499EA 1230 SGD
ICNA0855499
Anti-Ig Goat Antibody (FITC (Fluorescein Isothiocyanate))
Antibodies
The total protein is measured using the Biuret procedure with bovine albumin as the standard; F/P ratio is calculated using absorbance at 491.5nm. Antibody titer of the IgG fraction is standardized with an in-house control by immunoelectrophoresis; No trace of albumin is detected. Antibody activity to non-immunoglobulin serum proteins is not present.

Type: Secondary
Antigen: Ig
Clonality: Polyclonal
Clone:
Conjugation: FITC (Fluorescein Isothiocyanate)
Epitope:
Host: Goat
Isotype: IgG
Reactivity: Mouse
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR