Human IgM Isotype Control

Supplier: MP Biomedicals
0855916
ICNA0855916EA 1300 SGD
ICNA0855916
Human IgM Isotype Control
Antibodies
Product is human IgM and buffer salts.The product is tested for purity at a minimum of 20 mg/mL using immunoelectrophoresis. The product is human IgM; no trace of IgG, IgA or non-immunoglobulin serum proteins is detected. The total protein is determined using the Biuret procedure, with bovine albumin as the standard. Each vial is overfilled by approximately 10 percent so that a minimum of 5 mg of human IgM is recovered.
Order Now

SPECIFICATIONS

Environmentally Preferable
Antibody type Control
Conjugation Unconjugated
Reactivity
Host Human
Isotype IgM
Western blot Yes
ELISA Yes
Form Lyophilized
Purification Human IgM is purified from human myeloma serum using multi-step procedures which may include salt fractionation, gel filtration, ion-exchange chromatography and immunoabsorption. The product is dialyzed into 0.01M sodium phosphate, 0.07M sodium chloride, pH 7.3, filtered through a 0.22 μm filter and vialed. No preservative has been added.
Concentration Total Protein: 5.3 mg/mL; Volume: 19.4 mL
Shipping temperature RT

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR