Anti-beta-galactosidase Goat Polyclonal Antibody

Supplier: MP Biomedicals
0856030
ICNA0856030EA 685 SGD
ICNA0856030
Anti-beta-galactosidase Goat Polyclonal Antibody
Antibodies
  • Liquid form
  • There is no cross-reactivity to E.coli
  • Ideal for IFA, cell staining (fluorescent microscopy and cell sorting), tissue staining, and blot immunostaining

Type: Primary
Antigen: beta-galactosidase
Clonality: Polyclonal
Clone:
Conjugation: FITC
Epitope:
Host: Goat
Isotype: IgG
Reactivity:
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR