Anti-5-Dihydrotestosterone-15-CEMC-BSA Rabbit Polyclonal Antibody

Supplier: MP Biomedicals
08613401
ICNA08613401EA 130 SGD
ICNA08613401
Anti-5-Dihydrotestosterone-15-CEMC-BSA Rabbit Polyclonal Antibody
Antibodies
  • Working dilution: 1:100
  • Sensitivity: 10 pg/tube
  • Antibody affinity constant KA: 5,8 x 109 L/M (Scatchard Plot)

Type: Primary
Antigen: 5-Dihydrotestosterone-15-CEMC-BSA
Clonality: Polyclonal
Clone:
Conjugation:
Epitope:
Host: Rabbit
Isotype:
Reactivity: Human
Order Now

SPECIFICATIONS

Antigen symbol 5-DHT-15-CEMC-BSA
Antigen name 5-Dihydrotestosterone-15-CEMC-BSA
Environmentally Preferable
Antibody type Primary
Clonality Polyclonal
Reactivity Human
Host Rabbit
Isotype
Storage temperature 0 to +5 °C
Tested applications Radio Immunoassay

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR