Anti-Hemocyanin Rabbit Polyclonal Antibody

Supplier: MP Biomedicals
08618101
ICNA08618101EA 136 SGD
ICNA08618101
Anti-Hemocyanin Rabbit Polyclonal Antibody
Antibodies
It is a control reagent for the preparation and immunization of hapten-carrier conjugates.

Type: Primary
Antigen: Hemocyanin
Clonality: Polyclonal
Clone:
Conjugation:
Epitope:
Host: Rabbit
Isotype:
Reactivity:
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR