Streptomyces avidini Streptavidin, MP Biomedicals
Supplier: MP Biomedicals
08623001
ICNA08623001EA 474 SGD
ICNA08623001
Streptomyces avidini Streptavidin, MP Biomedicals
Proteins and Peptides
Streptavidin effectively binds to biotin with an isoelectric point close to neutral and it contains no carbohydrate residues.
Unit definition: One unit of streptavidin will bind 1 µg of biotin.