Anti-IgG PAN Mouse Monoclonal Antibody [clone: HP6017]

Supplier: MP Biomedicals
08630722
ICNA08630722EA 1230 SGD
ICNA08630722
Anti-IgG PAN Mouse Monoclonal Antibody [clone: HP6017]
Antibodies
This purified monoclonal antibody is supplied in 10 mM phosphate buffered saline, pH 7,4.

Type: Secondary
Antigen: IgG Fc PAN
Clonality: Monoclonal
Clone: HP6017
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG2ak
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR