Anti-IgE Mouse Monoclonal Antibody [clone: HP6029]

Supplier: MP Biomedicals
08634371
ICNA08634371EA 199 SGD
ICNA08634371
Anti-IgE Mouse Monoclonal Antibody [clone: HP6029]
Antibodies
This purified monoclonal antibody is supplied in 10 mM phosphate buffered saline, pH 7,4.

  • Dilution range: 10 µg/ml in PBS for EIA
  • Specificity: Human IgE heavy chain (Fc region)

Type: Secondary
Antigen: IgE Fc
Clonality: Monoclonal
Clone: HP6029
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG2ak
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR