Anti-Chondroitin-4-Sulphate Mouse Monoclonal Antibody [clone: BE-123]

Supplier: MP Biomedicals
08636511
ICNA08636511EA 1430 SGD
ICNA08636511
Anti-Chondroitin-4-Sulphate Mouse Monoclonal Antibody [clone: BE-123]
Antibodies
This mouse Anti-Chondroitin-4-Sulphate is used in ELISA, Radioimmunoassay, Western Blotting techniques and Immunohistochemistry.

  • Dilution range: 1:400 to 1:3200 for ELISA
  • Dilution range: 1:50 to 1:6400 for RIA
  • Dilution range: 1:1000 to 1:4000 for western blot

Type: Primary
Antigen: Chondroitin-4-Sulphate
Clonality: Monoclonal
Clone: BE-123
Conjugation:
Epitope:
Host: Mouse
Isotype: IgG1k
Reactivity: Bovine, Dog, Human, Mouse, Rat, Sheep
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR