Anti-Chondroitin-6-Sulphate Mouse Monoclonal Antibody [clone: MK302]

Supplier: MP Biomedicals
08636521
ICNA08636521EA 1080 SGD
ICNA08636521
Anti-Chondroitin-6-Sulphate Mouse Monoclonal Antibody [clone: MK302]
Antibodies
This mouse chondroitin 6 sulphate monoclonal antibody used in EIA, RIA and immunoblotting.

  • Dilution range: 1:100 to 1:5000 for ELISA
  • Dilution range: 1:100 to 1:200 for RIA

Type: Primary
Antigen: Chondroitin-6-Sulfate
Clonality: Monoclonal
Clone: MK302
Conjugation:
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity: Bovine, Dog, Human, Mouse, Rabbit, Rat, Sheep
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR