Bovine IgG Isotype Control

Supplier: MP Biomedicals
08641401
ICNA08641401EA 336 SGD
ICNA08641401
Bovine IgG Isotype Control
Antibodies
It is useful for structural and biological studies, as a reference standard, or as starting material for immunogen preparation. It is prepared using salt fractionation or by chromatographic techniques.

  • Immunoassay grade
  • Store at 2 to 8 °C
Order Now

SPECIFICATIONS

Antigen symbol IgG
Environmentally Preferable
Antibody type Control
Reactivity
Host Bovine
Isotype IgG
Storage temperature 2...8 °C

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR