Freund's complete adjuvant, MP Biomedicals
Supplier: MP Biomedicals
08642851
ICNA08642851EA 338 SGD
ICNA08642851
Freund's complete adjuvant, MP Biomedicals
Protein Expression Protein Expression Reagents
Freund's complete adjuvant is made by a modification of Freund's technique, in which 25 mg of Mycobacterium is suspended in a mixture of 7,5 ml Arlacel A and 42,5 ml paraffin oil.