Anti-IgG3 Sheep Polyclonal Antibody

Supplier: MP Biomedicals
08643121
ICNA08643121EA 377 SGD
ICNA08643121
Anti-IgG3 Sheep Polyclonal Antibody
Antibodies
Based on Immunoelectrophoresis, it produces precipitin band at a concentration of 1,0 mg/ml.

  • Can be reconstituted with 1,0 ml sterile distilled water
  • Dilution range: 1:20 for Immunoelectrophoresis

Type: Secondary
Antigen: IgG3
Clonality: Polyclonal
Clone:
Conjugation:
Epitope:
Host: Sheep
Isotype:
Reactivity: Human
Order Now

SPECIFICATIONS

Antigen symbol IgG3
Antigen name Immunoglobulin G3
Environmentally Preferable
Antibody type Secondary
Clonality Polyclonal
Reactivity Human
Host Sheep
Isotype
ImmunoChemistry Yes
Form Lyophilized
Storage temperature 0 to +5 °C
Tested applications Immunodiffusion, Immunoelectrophoresis

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR