Anti-IgE Human Polyclonal Antibody

Supplier: MP Biomedicals
08643491
ICNA08643491EA 840 SGD
ICNA08643491
Anti-IgE Human Polyclonal Antibody
Antibodies
By Immunoelectrophoresis, single arc was observed against antisera to whole human serum, human IgE and human kappa light chain.

  • Supplied in Tris or phosphate buffered saline

It is useful for structural and biological studies, as a reference standard, or as starting material for the preparation of an immunogen.

It is prepared from plasma shown to be nonreactive for HBsAg, anti-HCV, anti-HBc and negative for anti-HIV 1 and 2 by FDA-required tests.

Type:
Antigen: IgE
Clonality: Polyclonal
Clone:
Conjugation:
Epitope:
Host: Human
Isotype:
Reactivity:
Order Now

SPECIFICATIONS

Antigen symbol IgE
Antigen name Immunoglobulin E
Environmentally Preferable
Clonality Polyclonal
Reactivity
Host Human
Isotype
Storage temperature 0...5 °C
Tested applications Immunoelectrophoresis

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR