Anti-IgG, C3 IgG Polyclonal Antibody

Supplier: MP Biomedicals
08646381
ICNA08646381EA 694 SGD
ICNA08646381
Anti-IgG, C3 IgG Polyclonal Antibody
Antibodies
Type: 20269687
Antigen: Primary
Clonality: IgG, C3
Clone: Polyclonal
Conjugation:
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Cat
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR