Anti-Human Ferritin Rabbit Polyclonal Antibody, ImmunO™

Supplier: MP Biomedicals
08650771
ICNA08650771EA 285 SGD
ICNA08650771
Anti-Human Ferritin Rabbit Polyclonal Antibody, ImmunO™
Antibodies
Anti-Ferritin, Human is produced in rabbit using as immunogen ferritin isolated from normal human spleen.

Anti-ferritin, human antibody is suitable for use in Ouchterlony double diffusion, and immunoelectrophoresis. The antibody may also be used in western blot and immunoprecipitation.

Type: Primary
Antigen: Human-Ferritin
Clonality: Polyclonal
Clone:
Conjugation:
Epitope:
Host: Rabbit
Isotype:
Reactivity: Human
Order Now

SPECIFICATIONS

Environmentally Preferable
Antigen name Human- ferritin
Antibody type Primary
Clonality Polyclonal
Reactivity Human
Host Rabbit
Gene ID FTH1 2495
Isotype
Western blot Yes
ImmunoPrecipitation Yes
Immunogen Ferritin isolated from normal human spleen
Form Liquid
Concentration 7,61 mg/ml
Storage temperature +2 °C to +8 °C
Tested applications Immunoelectrophoresis

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR