Anti-Thyroxine BSA Serum Rabbit Polyclonal Antibody

Supplier: MP Biomedicals
08658501
ICNA08658501EA 1850 SGD
ICNA08658501
Anti-Thyroxine BSA Serum Rabbit Polyclonal Antibody
Antibodies
L-Thyroxine-BSA polyclonal antibody is used for radioimmunoassays.

  • Sensitivity: 1 ug thyroxine/tube
  • Working dilution: 1:1000 (using 0,1 ml/assay)
  • Can be reconstituted with 1,0 ml of distilled water

Specific activity for RIA system, second antibody and polyethylene glycol RIA method using 15 pg/tube iodine-125-thyroxine with specific activity of approximately 1000 uCi/ug as tracer and 0,1 ml of properly diluted antiserum.

Type: Primary
Antigen: Thyroxine BSA serum
Clonality: Polyclonal
Clone:
Conjugation:
Epitope:
Host: Rabbit
Isotype:
Reactivity:
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR