Anti-Human Properdin Goat Polyclonal Antibody

Supplier: MP Biomedicals
08680301
ICNA08680301EA 415 SGD
ICNA08680301
Anti-Human Properdin Goat Polyclonal Antibody
Antibodies
  • Dilution range: 1:4 for immunodiffusion

Type: Primary
Antigen: Human Properdin
Clonality: Polyclonal
Clone:
Conjugation:
Epitope:
Host: Goat
Isotype:
Reactivity: Human
Order Now

SPECIFICATIONS

Antigen name Properdin
Environmentally Preferable
Antibody type Primary
Clonality Polyclonal
Reactivity Human
Host Goat
Isotype
Form Liquid
Storage temperature –20 °C
Tested applications Ouchterlony Double Diffusion

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR