Anti-Bovine;Horse;Mouse Goat Polyclonal Antibody

Supplier: MP Biomedicals
08682651
ICNA08682651EA 492 SGD
ICNA08682651
Anti-Bovine;Horse;Mouse Goat Polyclonal Antibody
Antibodies
Goat anti-human secondary antibodies that are affinity-purified antibodies with well-characterized specificity for human immunoglobulins and are useful in the detection, sorting or purification of its specified target. Secondary antibodies offer increased versatility enabling users to use many detection systems (e.g. HRP, AP, fluorescence).

The antiserum contains 0,02 M sodium phosphate, 0,14 M sodium chloride, pH 7,3. No preservative is added. This reagent is the IgG fraction of the antiserum and was prepared by delipidation, ammonium sulfate fractionation and anion exchangeMore Product Information
Order Now

SPECIFICATIONS

Antigen symbol IgG
Environmentally Preferable
Antigen name Human serum IgG fraction
Antibody type Secondary
Clonality Polyclonal
Conjugation Unconjugated
Reactivity Human
Host Goat
Isotype IgG
ELISA Yes
ImmunoChemistry Yes
ImmunoPrecipitation Yes
ImmunoFluorescence Yes
Form Liquid
Purification The antiserum contains 0.02M sodium phosphate, 0.14M sodium chloride, pH 7.3. No preservative is added. This reagent is the IgG fraction of the antiserum and was prepared by delipidation, ammonium sulfate fractionation and anion exchange chromatography. The IgG content of the reagent is greater than 94% of total protein.
Concentration Protein Concentration: 36.5 mg/mL
Storage temperature +2 °C to +8 °C
Shipping temperature RT
Tested applications Immunoelectrophoresis
Electro Immunodiffusion
Immunofixation
Single Radial Immunodiffusion

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR