Anti-Actin Mouse Monoclonal Antibody [clone: B4]

Supplier: MP Biomedicals
08691331
ICNA08691331EA 715 SGD
ICNA08691331
Anti-Actin Mouse Monoclonal Antibody [clone: B4]
Antibodies
  • Dilution range: 1:200 to 1:500
  • Tissue source: skeletal muscle
  • Incubation time: 2 hours
  • Temperature: room temperature

Type: Primary
Antigen: Actin
Clonality: Monoclonal
Clone: B4
Conjugation:
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR