Rabbit Actin (from Muscle), MP Biomedicals
Supplier: MP Biomedicals
08771012
ICNA08771012EA 1100 SGD
ICNA08771012
Rabbit Actin (from Muscle), MP Biomedicals
Proteins and Peptides
Actin is a purified protein from Rabbit muscle, for use as a high molecular weight marker or as a standard.
It may be used as an antigen for antibody development.