Wheat germ agglutinin, MP Biomedicals
Supplier: MP Biomedicals
08790162
ICNA08790162EA 1440 SGD
ICNA08790162
Wheat germ agglutinin, MP Biomedicals
Proteins and Peptides
Agglutinin is isolated from wheat germ and purified by affinity chromatography.
It will agglutinate human red blood cells type A, B, AB, O and rabbit red blood cells.