Phytohemagglutinin (from Red kidney bean), MP Biomedicals
Supplier: MP Biomedicals
08791501
ICNA08791501EA 405 SGD
ICNA08791501
Phytohemagglutinin (from Red kidney bean), MP Biomedicals
Proteins and Peptides
Phytohemagglutinin PHA agglutinates the erythrocytes of all human blood groups as well as those of rabbit, dog, cat, mouse, sheep, and guinea pig.