Human native transferrin (from plasma), MP Biomedicals

Supplier: MP Biomedicals

08823431
ICNA08823431EA 1840 SGD
ICNA08823431
Human native transferrin (from plasma), MP Biomedicals
Proteins and Peptides

Transferrin is a glycoprotein with homologous N-terminal and C-terminal iron-binding domains. It is ideal for use as a growth factor for primary, continuous, and transformed cell cultures.


  • Soluble in 50 mg/ml purified water
  • No IgG's were detected
  • Endotoxin level: <1 EU/mg by LAL

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR