Cell detachment solution, Accutase, MP Biomedicals
Supplier: MP Biomedicals
091000449
ICNA091000449EA 221 SGD
ICNA091000449
Cell detachment solution, Accutase, MP Biomedicals
Cell Culture Supplements and Additives
Accutase solution is a trypsin replacement that contains both proteolytic and collagenolytic enzymes for cell detachment.
Accutase is effective in detaching primary fibroblasts, endothelial cells, neurons, tumour cell lines, and insect cells. It performs well in detaching cells for analysis of cell surface markers, virus growth assay, and flow cytometry as well as bioreactor scale-up.