Cell culture media, high growth enhancement medium, MP Biomedicals
Supplier: MP Biomedicals
091033822
ICNA091033822EA 243 SGD
ICNA091033822
Cell culture media, high growth enhancement medium, MP Biomedicals
Cell Culture Media
High Growth Enhancement Medium is a modification of DMEM in which 3,6 g/L fructose is substituted for glucose, resulting in better pH control and higher cell yields. Fructose is metabolised slower than glucose, resulting in slower acid build up. This media is recommended for the growth and support of cells on microcarrier matrices.