Cell culture media, liquid, RPMI 1640, MP Biomedicals
Supplier: MP Biomedicals
091646854
ICNA091646854EA 103 SGD
ICNA091646854
Cell culture media, liquid, RPMI 1640, MP Biomedicals
Cell Culture Media
RPMI 1640 medium was originally developed to culture human leukaemic cells in suspension and as a monolayer. Roswell Park Memorial Institute (RPMI) 1640 Medium has since been found suitable for a variety of mammalian cells, including HeLa, Jurkat, MCF-7, PC12, PBMC, astrocytes, and carcinomas. RPMI has successfully been used for the cultivation of normal human and neoplastic leukocytes. It is now a popular general purpose medium when properly supplemented.
  • A Dutch Modification of RPMI 1640 is also available through MP
  • Contains the reducing agent glutathione and high concentrations of vitamins
  • Used for the cultivation of normal human and neoplastic leukocytes
  • Popular general purpose medium when properly supplemented
  • Uses a sodium bicarbonate buffer system (2,0 g/L)
  • Requires a 5 to 10% CO2 environment to maintain physiological pH
RPMI 1640 medium is unique from other media because it contains the reducing agent glutathione and high concentrations of vitamins. It contains biotin, vitamin B12, and PABA, which are not found in Eagle's Minimal Essential Medium or Dulbecco's Modified Eagle Medium. In addition, the vitamins inositol and choline are present in very high concentrations. The medium contains no proteins, lipids, or growth factors. Therefore, it requires supplementation, commonly with 10% foetal bovine serum (FBS). It uses a sodium bicarbonate buffer system (2,0 g/L), and therefore requires a 5 to 10% CO₂ environment to maintain physiological pH.
RPMI 1640 Medium contains biotin, vitamin B12, and PABA, which are not found in Eagle's Minimal Essential Medium or Dulbecco's Modified Eagle Medium. In addition, the vitamins inositol and choline are present in very high concentrations. RPMI 1640 Medium contains no proteins, lipids, or growth factors. Therefore, RPMI 1640 Medium requires supplementation, commonly with 10% Fetal Bovine Serum (FBS).