Trace element solution 2, MP Biomedicals
Supplier: MP Biomedicals
091676649
ICNA091676649EA 231 SGD
ICNA091676649
Trace element solution 2, MP Biomedicals
Cell Culture Supplements and Additives
Trace Element Solution 2 is a 1000X sterile filtered solution, suitable for use with serum-free media; ideal for plant cell culture.
Trace Element Solution 2 is added aseptically to media as a supplement. Prior to use of the media it may be necessary to adjust pH and sterile filter (to reduce contamination).