Pig trypsin (from pancreas), MP Biomedicals

Supplier: MP Biomedicals

091689149
ICNA091689149EA 106 SGD
ICNA091689149
Pig trypsin (from pancreas), MP Biomedicals
Enzymes

Trypsin-EDTA solution is widely used for cell dissociation, routine cell culture passaging, and primary tissue dissociation due to its digestive strength.


  • Sterile
  • Concentration: 0.05% (w/v) Trypsin (1:250) and 0.02% (w/v) EDTA (Versene)
  • Formulation: EDTA 2Na 2H2O: 200.00000 mg/L, Potassium Chloride [KCl]: 400.00000 mg/L, Sodium Bicarbonate [NaHCO3]: 580.00000 mg/L, Sodium Chloride [NaCl]: 8000.00000 mg/L, Dextrose: 1000.00000 mg/L, Phenol Red Sodium Salt: 2.00000 mg/L, Trypsin 1:250: 500.00000 mg/L
  • pH: 7,0 to 7,4
  • Foreign Activity: Mycoplasma, Porcine Parvovirus : Not detected
  • Osmolality: 270 to 305 mM/kg
  • Storage and Handling: −20 °C


Trypsin-EDTA solution is widely used for cell dissociation, routine cell culture passaging, and primary tissue dissociation due to its digestive strength.

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR