TCM® Defined Serum Replacement, MP Biomedicals
Supplier: MP Biomedicals
092010026
ICNA092010026EA 555 SGD
ICNA092010026
TCM® Defined Serum Replacement, MP Biomedicals
Cell Culture Media
This serum replacement is effective in research situations such as studying the effects of growth factors or in the manufacturing of cell derived products.
  • A universal serum replacement that supports human and mammalian cells
  • Completely free of virus, mycoplasma and other infectious agents
  • Contains no growth factors, steroid hormones, extraneous proteins or phenol red
  • Protein origin - human; Drug Master on file with the FDA
  • Recommended for use in the production of cell-secreted proteins
TCM™ is a fortified, multipurpose serum replacement for long-term culturing of many types of anchorage dependent and suspension cultures. Highly effective in the production of biologicals, as well as in culturing other mammalian cells.