Multitest slides, MP Biomedicals
Supplier: MP Biomedicals
096041805 096041505 096041205
ICNA096041805EA 452 SGD
ICNA096041805 ICNA096041505 ICNA096041205
Multitest slides, MP Biomedicals
Slides Chamber Slides
Multitest slides for applications including immunofluorescence, cell culture, slide agglutination tests and histology.
  • Suitable for one-time use
  • Fragile glass with blue coating
  • Uncoated areas with wettable surface
For cell attachment and growth, the slides require a simple washing in deionised water or detergent, followed by sterilisation at 160 °C for one hour. They can resist to direct heat as well as common organic solvents and fixatives.
A four-channel pipette needs to be used to transfer samples from the microplate to the slide.
Order Now
SPECIFICATIONS
Colour Blue
Environmentally Preferable
W×D×H 25×75×1 mm