Acetate plate seal, MP Biomedicals
Supplier: MP Biomedicals
097640105
ICNA097640105EA 132 SGD
ICNA097640105
Acetate plate seal, MP Biomedicals
Seals Microplate Seals
Clear adhesive seals for multiwell plates and tissue culture plates.
  • Easy to apply
  • Suitable for both short and long term storage
  • Provides tight seal to minimise evaporation and condensation
Order Now
SPECIFICATIONS
Environmentally Preferable