DNA purification kits, GeneClean® for Ancient DNA™ Kit, MP Biomedicals
Supplier: MP Biomedicals
111002200
ICNA111002200EA 962 SGD
ICNA111002200
DNA purification kits, GeneClean® for Ancient DNA™ Kit, MP Biomedicals
Nucleic Acid Reagents Nucleic Acid Purification Kits and Reagents
The GeneClean® for Ancient DNA™ Kit is ideal for purification of fragmented or damaged DNA from aged or preserved samples. It is designed for isolation of up to 20 μg DNA from bone, preserved tissue, soil organisms, or animal by-products.
GeneClean® for Ancient DNA™ Kit minimises the possibility of contamination by contemporary DNA from extraneous sources. All solutions are prepared from ultra-pure reagents and dispensed in a dedicated laminar flow hood.