Plasmid purification, MiniPrep Express™ Matrix, MP Biomedicals
Supplier: MP Biomedicals
112000200
ICNA112000200EA 1200 SGD
ICNA112000200
Plasmid purification, MiniPrep Express™ Matrix, MP Biomedicals
Nucleic Acid Reagents Nucleic Acid Purification Kits and Reagents
MiniPrep Express™ Matrix is designed for rapid purification of plasmid DNA. It is an ideal tool for screening hundreds of recombinant clones in record time using restriction enzyme analysis and PCR.
MiniPrep Express™ Matrix provides the essential ingredients for 'make your own' miniprep kits for rapid screening that involve a restriction digest. This kit allows you to make the lysis reagents required for the most commonly used methods.
Delivery information: Kit contains MiniPrep Express™ Matrix, protocols and recipes necessary for either alkaline lysis or boiling miniprep procedures.