Dehydrated culture media, LB-medium, molecular biology grade, MP Biomedicals
Supplier: MP Biomedicals
113002216 113002046 113002236 113002232 113002231 113002226 113002222 113002221 113002122 113002332 113002042 113002041 113002031 113002026 113002022 113002021 113002241 113002331 113002011 113002016 113002056 113002121 113002131 113002132 113002136 113002141 113002146 113002211 113002242 113002246
ICNA113002216EA 205 SGD
ICNA113002216 ICNA113002046 ICNA113002236 ICNA113002232 ICNA113002231 ICNA113002226 ICNA113002222 ICNA113002221 ICNA113002122 ICNA113002332 ICNA113002042 ICNA113002041 ICNA113002031 ICNA113002026 ICNA113002022 ICNA113002021 ICNA113002241 ICNA113002331 ICNA113002011 ICNA113002016 ICNA113002056 ICNA113002121 ICNA113002131 ICNA113002132 ICNA113002136 ICNA113002141 ICNA113002146 ICNA113002211 ICNA113002242 ICNA113002246
Dehydrated culture media, LB-medium, molecular biology grade, MP Biomedicals
Media for Microbiology Dehydrated Media for Microbiology
Rich medium for the growth of E. coli and certified for plasmid and other bacterial cultures in liquid medium.
  • Proprietary media for enhanced cell yield
  • Convenient packaging formats for a variety of applications and budgets
  • Proven and reliable reputation
  • High-quality raw materials and additives
  • Encapsulated media
  • No weighing, dust and cleanup
  • Drop capsules into water and autoclave
LB agar medium is a nutritionally rich medium originally developed for growth and maintenance of E. coli strains. The ultimate in ease and convenience, LB agar medium in a capsule form eliminates the waste, inaccuracies, and mess associated with weighing out bulk powder.The growth characteristics are the same as LB agar powder. LB agar medium supports a luxurious growth of E. coli cells as tryptone and yeast extract supply essential growth factors that E. coli cells would otherwise have to synthesise. The medium also contains essential electrolytes for transport and osmotic balance, due to the NaCl component.
Application includes: Bacteria cell culture, gene expression, transformation, microbiological analysis, protein expression.
Certifications: Molecular Biology Certified™. Gelatin capsules are produced in a facility that meets the FDA’s recent guidance as being derived from BSE-free herds that are born, raised, and slaughtered in the United States.
Order Now
SPECIFICATIONS
Environmentally Preferable
Storage temperature Store sterile LB Agar Medium at ambient temperature (15 - 30 °C). Powder medium is hygroscopic, therefore minimize exposure to air to prolong shelf life of media RT Store sterile LB Agar Medium at ambient temperature (15 - 30 °C). Powder medium is hygroscopic, therefore minimize exposure to air to prolong shelf life of media.