Dehydrated culture media, SC-Ura media, MP Biomedicals
Supplier: MP Biomedicals
114410622
ICNA114410622EA 1410 SGD
ICNA114410622
Dehydrated culture media, SC-Ura media, MP Biomedicals
Media for Microbiology Dehydrated Media for Microbiology
SC-Ura is a single dropout formulation of Synthetic Complete Supplement Mixture (SC) of amino acids for S. cerevisiae growth media.
  • Create a richer medium for better yield and growth rate
  • Increase the probability of successful transformations when screening libraries or performing gene knock-outs
Also known as Hopkins mixture and is available in complete and dropout formulations.