DNA extraction kits, tissue DNA extraction kit, MPure™, MP Biomedicals
Supplier: MP Biomedicals
117022400
ICNA117022400EA 1070 SGD
ICNA117022400
DNA extraction kits, tissue DNA extraction kit, MPure™, MP Biomedicals
Nucleic Acid Reagents Nucleic Acid Purification Kits and Reagents
MPure™ Tissue DNA Extraction Kit is used with the MPure-12™ Automated Nucleic Acid Purification System for extraction and purification of genomic DNA from a variety of animal tissues, swab samples and blood stains. Nucleic acids extracted and purified from MPure™ Tissue DNA Extraction Kits can be used in a number of downstream applications including: PCR, qPCR, Sequencing (NGS), Microarray, RFLP and Southern Blot Analysis.