Handheld digital microscopes, Dino-Lite High Magnification
Supplier: Dino-Lite / IDCP B.V.
AM4113T5
IDCPAM4113T5EA 1210 SGD
IDCPAM4113T5
Handheld digital microscopes, Dino-Lite High Magnification
Microscopes
Dino-Lite models within the High Magnification series exceed a magnification of 200 times and offer magnifications of 400×, 500× or even 900 times. Magnification range: 400 - 900×.
  • High speed USB 2.0 connection
  • Metal housing or composite housing
  • Edge series models with high-quality optics provide a very sharp, bright and natural colour image with very low aberration and vignetting
  • Wireless streaming to a tablet, smartphone, or computer is possible in combination with WF-10 unit (to be ordered separately)
  • Integrated polariser greatly reduces glare on
More Product Information
Order Now
SPECIFICATIONS
Environmentally Preferable