Rotary Evaporators
Supplier: IKA
10004800 10004803 10004806
IKAA10004800EA 11730 SGD
IKAA10004800 IKAA10004803 IKAA10004806
Rotary Evaporators
Evaporators Rotary Evaporators
Rotary evaporators with integrated heating bath, safe and simple operation via ergonomic controls situated at the front.
  • All models have motorised lift with safety stop function
  • Digital speed and digital bath temperature display
  • Infrared interface transfers data from heating bath to the drive unit
  • Digital heating bath with microprocessor controller can be used with oil or water, features integrated carrying handles for increased safety
  • RS232 interface for remote PC operation using labworldsoft® software (optional), to allow for operation in automatic mode
More Product Information