Magnetic stirring bars, PTFE
Supplier: IKA
1358600
IKAA1358600EA 387 SGD
IKAA1358600
Magnetic stirring bars, PTFE
Stirrers Magnetic Stirring Bars
  • Very good chemical resistance
  • Temperature resistant, suitable for high and low temperature applications