Homogenisers, T 25 digital LR
Supplier: IKA
20010537
IKAA20010537EA 5000 SGD
IKAA20010537
Homogenisers, T 25 digital LR
Homogenisers Homogenisers Bench
Powerful disperser for volumes ranging from 1 to 2000 ml (H2O) with digital speed display. The wide speed range of 600 to 25000 rpm facilitates work with high circumferential speeds. An extensive selection of dispersing shafts makes multiple applications possible. The application range includes homogenisation and dispersion tasks for use under vacuum in the IKA laboratory reactors.
  • Digital speed display
  • Electronic speed control
  • Electronic overload protection
  • Quick and easy cleaning of stainless steel dispersing shafts
  • Error code display
  • Quiet operation
More Product Information